Learn More
SPEX CERTIPREP LLC 3P ICP-MS DAILY CHECK, 250 ML
Catalog No. NC1743537
Encompass_Preferred
(HPS by Zeptometrix) 10 Element ICPMS Stock Tuning Solution: Beryllium, Cerium, Cobalt, Indium, Iron, Lead, Magnesium, Thorium, and Uranium - 1 g/mL, Barium - 10 g/mL in 2% HNO3
Catalog No. NC1743537 Supplier SPEX CERTIPREP LLC 3P Supplier No. ICPMSTS15250
(HPS by Zeptometrix) 10 Element ICPMS Stock Tuning Solution: Beryllium, Cerium, Cobalt, Indium, Iron, Lead, Magnesium, Thorium, and Uranium - 1 g/mL, Barium - 10 g/mL in 2% HNO3