Learn More
Inorganic Ventures ICP MS INTERNAL STD 125mL
SDP
Catalog No. 50723667
Encompass_Preferred
Matrix: Nitric Acid (HNO3), Analyte: Bi, Tb, 6Li, In, Sc, and Y 10 ug/mL each.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-667 Supplier Inorganic Ventures Supplier No. IVICPMS71D125ML
Matrix: Nitric Acid (HNO3), Analyte: Bi, Tb, 6Li, In, Sc, and Y 10 ug/mL each.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search