Learn More
Inorganic Ventures AGILENT TUNING SOLUTION 125mL
SDP
Catalog No. 50723169
Encompass_Preferred
Matrix: Nitric Acid (HNO3), Instrument(s): Agilent/Varian, Analyte: Ce, Li, Y, Co, and Tl 10 ug/mL each.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-169 Supplier Inorganic Ventures Supplier No. AGITS1125ML
Matrix: Nitric Acid (HNO3), Instrument(s): Agilent/Varian, Analyte: Ce, Li, Y, Co, and Tl 10 ug/mL each.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search