Learn More
Agilent Technologies Tuning Solution For ICP-MS, Inorganic, 2-Piece Set
Catalog No. NC1625063
Encompass
The Tuning Solution for ICP-MS is a multi-element standard designed for tuning and performance checking of Agilent 7500 series ICP-MS systems. This ready-to-use premixed kit contains lithium (Li), magnesium (Mg), yttrium (Y), cerium (Ce), thallium (Tl), and cobalt (Co) at a concentration of 1 μg/L each in a 2% nitric acid matrix. Each standard is produced gravimetrically and volumetrically using high-purity starting materials and NIST-traceable standards to ensure accuracy and stability.
  • Contains six elements (Li, Mg, Y, Ce, Tl, Co) in a 2% HNO3 matrix
  • Pre-mixed, ready-to-use formulation reduces preparation time and contamination risk
  • Quality controlled using ICP/ICP-MS spectrometry via NIST SRMs
  • Packaged in acid-leached, triple-rinsed bottles
  • Certified accuracy of ±10% of measured analyte concentration
Catalog No. NC1625063 Supplier Agilent Technologies Supplier No. 51855959
The Tuning Solution for ICP-MS is a multi-element standard designed for tuning and performance checking of Agilent 7500 series ICP-MS systems. This ready-to-use premixed kit contains lithium (Li), magnesium (Mg), yttrium (Y), cerium (Ce), thallium (Tl), and cobalt (Co) at a concentration of 1 μg/L each in a 2% nitric acid matrix. Each standard is produced gravimetrically and volumetrically using high-purity starting materials and NIST-traceable standards to ensure accuracy and stability.
  • Contains six elements (Li, Mg, Y, Ce, Tl, Co) in a 2% HNO3 matrix
  • Pre-mixed, ready-to-use formulation reduces preparation time and contamination risk
  • Quality controlled using ICP/ICP-MS spectrometry via NIST SRMs
  • Packaged in acid-leached, triple-rinsed bottles
  • Certified accuracy of ±10% of measured analyte concentration