Learn More
Neta Scientific ICP-OES Wavelength Calibration Solution, 500 mL, 5% HNO3, Multi-element, Al, As, Ba, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Se, Sr, Zn 5 mg/L, K 50 mg/L
SDP
Catalog No. NC0830447
Encompass_Preferred
This item is manufactured by Agilent
ICP-OES Wavelength Calibration Solution, 500 mL, 5% HNO3, Multi-element, Al, As, Ba, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Se, Sr, Zn 5 mg/L, K 50 mg/L
Catalog No. NC0830447 Supplier Neta Scientific Supplier No. 6610030100
This item is manufactured by Agilent
ICP-OES Wavelength Calibration Solution, 500 mL, 5% HNO3, Multi-element, Al, As, Ba, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Se, Sr, Zn 5 mg/L, K 50 mg/L