Learn More
Thermo Scientific ICP PUMP TUBING-INT STD KIT
Supplier:  Thermo Scientific 842312051551
Encompass
Internal Standards Kit; Internal standards mixing kit, comprising 6 Tygon orange/white pump; tubes, 6 Tygon blue/white pump tubes, Y-piece, mixing loop and; connecting tube for fitting.; Compatible with all iCAP 6000 Series Spectrometers.
Catalog No. NC9973085