Learn More
AccuStandard, Inc ICP QUALT. CONTR. STD NO1
SDP
Catalog No. NC0274630
Encompass
ICP Quality Control Standard #1. 100 mL. 10 ?g/mL in 2-5% Nitric Acid.
Catalog No. NC0274630 Supplier AccuStandard, Inc Supplier No. QCS011
ICP Quality Control Standard #1. 100 mL. 10 ?g/mL in 2-5% Nitric Acid.