Learn More
Thermo Scientific HR-ICP-MS System Injectors | | 1 EA
Supplier:  Thermo Scientific 1122800
Encompass
Tailor your consumables to your chosen sample matrices for enhanced analysis,This wide selection of reliable easy to install Injectors with differing internal diameters and materials ensures that your system is optimized precisely to meet your analytical requirements,All our injectors are easily removable for improved robustness and reduced maintenance requirements
Catalog No. NC0312212