Learn More
Agilent Technologies SPS 4 probe 0.8 mm id with green markers
Catalog No. NC1421372
Encompass
SPS 4 probe 0.8 mm id with green markers. Inert, FEP sleeved for SPS 4 autosampler.
Catalog No. NC1421372 Supplier Agilent Technologies Supplier No. G841080102
SPS 4 probe 0.8 mm id with green markers. Inert, FEP sleeved for SPS 4 autosampler.