Learn More
ICP Solvent, For inorganic trace analysis, 99.9995% (metals basis), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| CAS | 64742-47-8 |
| Melting Point | -58°C |
| Density | 0.810-0.838 g/mL (at 20°C) |
| Boiling Point | 200°C to 250°C |
| Quantity | 1 L |
| Synonym | Hydrotreated light distillate (petroleum) |
| Percent Purity | ≥99.9995% (metals basis) |
| Grade | For inorganic trace analysis |
| Physical Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.