Learn More
Cayman Chemical ICR-1 (potassium salt), Purity: ≥90%, 0.5 mg
SDP
Catalog No. 502913171
Encompass_Preferred
A cell-impermeable fluorescent calcium indicator; binds to calcium (Kd = 480 mM); ex/em = 580/660 nm, respectively; can be used in lipid membrane-free systems or liposomes or can be introduced into cells by electroporation, microinjection, or other methods
Catalog No. 50-291-3171 Supplier Cayman Chemical Supplier No. 349860.5 MG
A cell-impermeable fluorescent calcium indicator; binds to calcium (Kd = 480 mM); ex/em = 580/660 nm, respectively; can be used in lipid membrane-free systems or liposomes or can be introduced into cells by electroporation, microinjection, or other methods