Learn More
IPHASE PHARMA SERVICES LLC ICR/CD-1 Mouse PBMC,Frozen,5million
Catalog No. NC3967049
Encompass_Preferred
Peripheral Blood Mononuclear Cells (PBMCs) are a heterogeneous population of immune cells found in peripheral blood and isolated via density gradient centrifugation. They include lymphocytes—such as T cells, B cells, and natural killer (NK) cells—and monocytes.It can be applicated Immunological Research and Cell Therapy (CAR-T/CAR-NK). These products require shipment in LN2 container therefore customer need to return the container.
Catalog No. NC3967049 Supplier IPHASE PHARMA SERVICES LLC Supplier No. 082E01.21
Peripheral Blood Mononuclear Cells (PBMCs) are a heterogeneous population of immune cells found in peripheral blood and isolated via density gradient centrifugation. They include lymphocytes—such as T cells, B cells, and natural killer (NK) cells—and monocytes.It can be applicated Immunological Research and Cell Therapy (CAR-T/CAR-NK). These products require shipment in LN2 container therefore customer need to return the container.