Learn More
Medchemexpress LLC Icrf-193, apoptosis inducer, arabinosidase substrate | 21416-88-6 | MFCD01702964 | 99.6% | 282.30 | C12H18N4O4 | 100 MG
SDP
Catalog No. 5000228925
Encompass_Preferred
ICRF-193 is a DNA topoisomerase II inhibitor provided as a white to off-white solid for research use only. It inhibits DNA synthesis and induces apoptosis, and is used in studies of cancer, inflammation, and cardiovascular biology.
  • DNA topoisomerase II inhibitor used in cellular and molecular studies.
  • White to off-white solid suitable for preparing stock solutions.
  • High purity for reproducible experimental results.
  • Stable when stored at -20°C; protect from light.
  • Molecular weight 282.30 g/mol; formula C12H18N4O4.
Catalog No. 50-002-28925 Supplier Medchemexpress LLC Supplier No. HY118590100MG
Add to Cart
Add to Cart
Request a Quote
ICRF-193 is a DNA topoisomerase II inhibitor provided as a white to off-white solid for research use only. It inhibits DNA synthesis and induces apoptosis, and is used in studies of cancer, inflammation, and cardiovascular biology.
  • DNA topoisomerase II inhibitor used in cellular and molecular studies.
  • White to off-white solid suitable for preparing stock solutions.
  • High purity for reproducible experimental results.
  • Stable when stored at -20°C; protect from light.
  • Molecular weight 282.30 g/mol; formula C12H18N4O4.