Learn More
Selleck Chemical LLC iCRT3 5mg 901751-47-1
SDP
Supplier:  Selleck Chemical LLC S86475MG
Encompass_Preferred
iCRT3 is an antagonist of Wnt/?-catenin signaling with an IC50 of 8.2 nM in the Wnt responsive STF16-luc reporter assays. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1901474