Learn More
Medchemexpress LLC HY-103705 5mg Medchemexpress, iCRT3 CAS:901751-47-1 Purity:>98%
SDP
Catalog No. 501968167
Encompass_Preferred
Medchemexpress, HY-103705 5mg iCRT3 CAS:901751-47-1 iCRT3 is an inhibitor of both Wnt and β-catenin-responsive transcription. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8167 Supplier Medchemexpress LLC Supplier No. HY1037055MG
Medchemexpress, HY-103705 5mg iCRT3 CAS:901751-47-1 iCRT3 is an inhibitor of both Wnt and β-catenin-responsive transcription. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.