Learn More
Andwin Scientific ICY BLUE 1L SM ICE PAN EA
SDP
Catalog No. 50727420
Encompass_Preferred
The Glacierbrand Small Ice Pan features thick, polystyrene insulated walls leaving no puddles of condensation. These are also stackable, lightweight and chemically resistant. It is in Icy Blue color, sold per piece.
Catalog No. 50-727-420 Supplier Andwin Scientific Supplier No. 633111S
The Glacierbrand Small Ice Pan features thick, polystyrene insulated walls leaving no puddles of condensation. These are also stackable, lightweight and chemically resistant. It is in Icy Blue color, sold per piece.