Learn More
Andwin Scientific ICY BLUE 4L MD ICE PAN EA
SDP
Catalog No. 50727419
Encompass_Preferred
The Glacierbrand Medium Ice Pan features thick, polystyrene insulated walls leaving no puddles of condensation. These are also stackable, lightweight and chemically resistant. It is in Icy Blue color, sold per piece.
Catalog No. 50-727-419 Supplier Andwin Scientific Supplier No. 633111M
The Glacierbrand Medium Ice Pan features thick, polystyrene insulated walls leaving no puddles of condensation. These are also stackable, lightweight and chemically resistant. It is in Icy Blue color, sold per piece.