Learn More
Andwin Scientific ICY BLUE 9L LG ICE PAN EA
SDP
Catalog No. 50727418
Encompass_Preferred
The Glacierbrand Large Ice Pan features thick, polystyrene insulated walls leaving no puddles of condensation. These are also stackable, lightweight and chemically resistant. It is in Icy Blue color, sold per piece.
Catalog No. 50-727-418 Supplier Andwin Scientific Supplier No. 633111L
The Glacierbrand Large Ice Pan features thick, polystyrene insulated walls leaving no puddles of condensation. These are also stackable, lightweight and chemically resistant. It is in Icy Blue color, sold per piece.