Learn More
Miele, Inc. ID 180 INJECTOR
Catalog No. NC2233919
Encompass_Preferred
Injector nozzle with plastic supports, Ø 0 3/16 (4), length 180 mm.Suitable for standard mobile injector units and injector basketsSuitable for measuring flasks and test tubes. MIELE INC mfg# 03810360
This item is not returnable. View return policy
Catalog No. NC2233919 Supplier Miele, Inc. Supplier No. 03810360
Injector nozzle with plastic supports, Ø 0 3/16 (4), length 180 mm.Suitable for standard mobile injector units and injector basketsSuitable for measuring flasks and test tubes. MIELE INC mfg# 03810360