Learn More
Selleck Chemical LLC ID-8
SDP
Supplier:  Selleck Chemical LLC S732750MG
Encompass_Preferred
ID-8 is a DYRK inhibitor and sustains embryonic stem cell self-renewal in long-term culture
Catalog No. 50-000-04406