Learn More
AdipoGen ID-8 (5 mg)
SDP
Supplier:  AdipoGen AGCR13655M005
Encompass_Preferred
Chemical. CAS: 147591-46-6. Formula: C16H14N2O4. MW: 298.3. Synthetic. Cell culture supplement that can sustain self-renewal and pluripotency of mouse embryonic stem cells (ESCs) in vitro. At 10µM it stimulates proliferation in serum-free media at a steady rate for more than 30 days. Reversibly maintains the long-term culture of ESCs by sustaining Nanog gene expression through Sox2-Oct3/4 activation and enhances Wnt-mediated hESC survival and proliferation via inhibition of DYRKs (DYRK2 and DYRK4).
Catalog No. 50-205-4032