Learn More
Med Vet International KVP I.D. Band, 10in, 500/Box
Catalog No. 502830490
Encompass_Preferred
KVP 10in. ID band is manufactured with strong and tough spun bonded olefin. They have a strong, pressure sensitive adhesive that becomes more permanent with increased pressure. You can use permanent markers, ball point pens, and pencils to mark these ID Bands. This product contains 500, mixed colored ID Bands.
Catalog No. 50-283-0490 Supplier Med Vet International Supplier No. EJIDM10
KVP 10in. ID band is manufactured with strong and tough spun bonded olefin. They have a strong, pressure sensitive adhesive that becomes more permanent with increased pressure. You can use permanent markers, ball point pens, and pencils to mark these ID Bands. This product contains 500, mixed colored ID Bands.