Description
QuidelOrtho ID-Micro Typing System is suitable for a broad range of blood bank applications, including antibody screening and identification, ABO blood grouping and Rh phenotyping, compatibility testing, and reverse serum grouping and antigen typing, all using proven serologic methods. Gel Test cards are prefilled cards containing gel mixed with the appropriate reagent. Simply pipet, incubate, spin and read: results are ready to report, without the need for cell washing, red cell resuspension or tube shaking.
Standardization
- Standardized methods ensure uniform results from tech-to-tech and laboratory-to-laboratory
- Enhanced cGMP compliance is attained through standardized procedures and simplified training
Safety
- Opportunities for error are reduced by stable and clear endpoints that provide objective, consistent results
- Ease of Use
- Gel testing is easy to learn, use, and teach: the ideal technology for all shifts and all levels of blood banking experience
- Eliminates washing, shaking, and the need to add control cells
Cost-effectiveness
- Gel testing requires fewer steps than tube testing, dramatically reducing hands-on time, with less waste and costly re-work
- Well-balanced sensitivity and specificity means less time and fewer resources spent on clinically unimportant antibodies
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Certifications/Compliance | cGMP |
| Type | ID-Micro Typing System™ A/B/D Monoclonal and Reverse Grouping Card |
| For Use With (Application) | For broad range of Blood bank applications, Antibody screening and identification, ABO Blood grouping and Rh phenotyping, compatibility testing and reverse Serum grouping and Antigen typing |
| Includes | Gel-filled microtubes |
| Quantity | 100 Cards |