Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuidelOrtho ID-MTS™ Daily QC Simulated Whole Blood Quality Control Kit
SDP

Catalog No. 23900713 Shop All Ortho-Clinical Diagnostics Products
Change view
Click to view available options
Form:
Liquid Ready-to-Use
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
23-900-713 Liquid Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-900-713 Supplier QuidelOrtho Supplier No. 6708858
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The QuidelOrtho Daily QC kit enables labs to test QC with a standardized workflow for the wide-ranging menu on the QuidelOrtho Vision Swift Platform.

Streamline your QC testing with the same trusted results from the Ortho Vision™ Swift Platform.

As part of our QuidelOrtho Vision Swift Platform:

  • New Rh phenotyping capability for ID-Micro Typing System™ (ID-MTS) Gel Cards: Quality control (QC) for Rh phenotyping just got easier. Eliminate the need to create user-defined samples, saving you valuable time.
  • Longer open vial stability and shelf life: Open vial stability has increased from seven to 14 days, and shelf life has been extended by up to 27 days.

With the same standardized approach and workflow to QC across a wide-ranging menu, your new QuidelOrtho Daily QC kit ensures the pre-transfusion testing you perform each day will continue to deliver the right results and put patient safety first.

  • Vial 1 - Group A1 B, DCEce
  • Vial 2 - Group O, DCe containing Anti-A, Anti-B and Anti-c
  • Vial 3 - Group A, DcE containing Anti-B and Anti-Fya
  • Vial 4 - Group B, ce, containing Anti-A and Anti-D (concentration of approximately 0.1 IU/mL)

Specifications

Content And Storage Store at 2°C to 8°C
CE Marker Yes
Type Quality Control Kit
For Use With (Application) Qualitative controls of ABO and Rh phenotyping and antibody screening
For Use With (Equipment) QuidelOrtho Workstation with or without QuidelOrtho Optix™ Reader, QuidelOrtho VISION™ Analyzer, and QuidelOrtho VISION™ Max Analyzer
Dimensions (L x W x H) 53 x 53 x 107 mm
Form Liquid Ready-to-Use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.