Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ ID NOW™ COVID-19 2.0 24 Test Kit PROMO

Catalog No. 23046655PM Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-046-655PM 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-046-655PM Supplier Abbott™ Supplier No. 192000P
Get 6 Test Kits for the Price of 5
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

The ID NOW COVID-19 2.0 24 Test Kit helps clinicians to make faster decisions with diagnostic results in real time. Buy 5, get 6.

  • CLIA-waived ID NOW™ COVID-19 2.0 assay delivers reliable molecular results in just minutes
  • Gives real-time access to the information you need to make actionable decisions at the point of care
  • Uses direct nasal or nasopharyngeal swabs
  • Assay kit can be stored at room temperature and contains 24 tests, nasal swabs, and a positive control swab
  • ID NOW Platform features bidirectional connectivity to support easy data integration from multiple instruments to hospital/lab information system.
  • Positive results as early as 6 minutes, negative results in 12 minutes
  • Nucleic Acid Amplification Technology (NAAT), providing molecular results in a fraction of the time it takes traditional lab tests
  • Requires minimum training with easy on-screen steps to guide users
  • You can decide which tests to run based on patient presentation, circulating prevalence and seasonality of infectious diseases

Specifications

Certifications/Compliance FDA 510(k)
CE Marker No
Clia Complexity Waived (FDA, EUA)
Detectable Analytes SARS-CoV-2
Sample Type Nasal Swab, Nasopharyngeal Swab
Type COVID-19 Test Kit
No. of Reactions 24 Tests
Quantity 1 Kit
Dimensions (L x W x H) 207 x 194 x 145 mm (8.2 x 7.6 x 5.7 in.)
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.