Learn More
Abbott ID NOW™ Rapid Molecular Instrument and ID NOW™ Rapid Molecular RSV Test Kits PROMO
Catalog No. NC3432935 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Type:
Rapid Molecular RSV Test Kits
Catalog No. Type
NC3432935 Rapid Molecular RSV Test Kits
1 options
This item is not returnable. View return policy
Catalog No. NC3432935 Supplier Abbott Supplier No. IDNOW7R
Add to Cart
Add to Cart
This item is not returnable. View return policy
Promotional Kit that contains one ID NOW™ Rapid Molecular Instrument and seven ID NOW™ Rapid Molecular RSV Test Kits.
For a limited time, Expand your testing capacity with these ID NOW™ Promotions:
  • The CLIA-waived ID NOW™ Rapid Molecular Platform provides molecular results within 2 to 13 minutes, maximizing patient care efficiencies and simplifying workflow with on-screen steps to guide users and no manual pipetting required.
  • Promotion Item IDNOW7R contains one (1) ID NOW™ Rapid Molecular INSTRUMENT and seven (7) ID NOW™ Rapid Molecular RSV Test Kits.
Specifications
Detectable Analytes Respiratory Syncytial Virus
Type Rapid Molecular RSV Test Kits