Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Abbott ID NOW™ Rapid Molecular Instrument and ID NOW™ Rapid Molecular Strep A Test Kits PROMO
Catalog No. NC3307569 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Detectable Analytes:
Streptococci (Group A)
Catalog No. Detectable Analytes
NC3307569 Streptococci (Group A)
1 options
Catalog No. NC3307569 Supplier Abbott Supplier No. IDNOW8S
Add to Cart
Add to Cart
Promotional Kit that contains one ID NOW™ Rapid Molecular Instrument and eight ID NOW™ Rapid Molecular Strep A Test Kits.
For a limited time, Expand your testing capacity with these ID NOW™ Promotions:
  • The CLIA-waived ID NOW™ Rapid Molecular Platform provides molecular results within 2 to 13 minutes, maximizing patient care efficiencies and simplifying workflow with on-screen steps to guide users and no manual pipetting required.
  • Promotion Item IDNOW8S contains one (1) ID NOW™ Rapid Molecular INSTRUMENT and eight (8) ID NOW™ Rapid Molecular Strep A Test Kits.
Specifications
Detectable Analytes Streptococci (Group A)
Type ID NOW™ Rapid Molecular Instrument and Strep A Test Kits