Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ ID NOW™ Strep A 2 Test Kit PROMO

Catalog No. 23046632PM Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Detectable Analytes:
Streptococci (Group A)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
23-046-632PM Streptococci (Group A)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-046-632PM Supplier Abbott™ Supplier No. 734000P

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The fastest CLIA-waived Strep A Molecular Test - positive results in a few as 2 minutes. Buy 5, get 6.

Abbott ID NOW Strep A 2 is now the fastest Strep A test without the need for culture confirmation. It provides positive results in as little as 2 minutes; allowing you to confidently test, and with a positive result, treat during the patient visit. Faster than other molecular methods and more accurate than conventional rapid tests, Abbott ID NOW Strep A 2 delivers results within an actionable time frame allowing you to make real-time clinical decisions which impact patient care.
  • Now CLIA-waived
  • Offers exceptional performance
  • Positive results in as little as 2 minutes
  • Nearly 90% fewer false negatives than RADTs
  • Enhanced molecular sensitivity
  • Eliminates the need for culture confirmation of negative results
  • Allows for the accurate detection of Group A Strep
  • Increased detection compared to culture
  • Eliminates interpretation and transcription errors
  • Gives you the confidence to make clinical decisions sooner
  • Facilitates effective patient management
  • Small footprint enables ease of use in any healthcare setting
  • Aids targeted antiviral therapy and Antimicrobial Stewardship
  • Specimen type: throat swab

Specifications

Description Strep A Test Kit
Detectable Analytes Streptococci (Group A)
Sample Type Throat Swab
Type Strep A Test
For Use With (Application) For accurate detection of Group A Strep
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.