Learn More
Aviva Systems Biology Corp ID3 Antibody - N-terminal region (ARP32335_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32335_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ID3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ID3. Alternate names: HEIR-1, bHLHb25ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-54530