Learn More
Molecular Devices Molecular Devices Injector System, SpectraMax iD3 Compatible, Two Injectors
Catalog No. 5000014673
Encompass_Preferred
The SpectraMax iD3 Injector System adds automated reagent injection capability to iD3-series multi-mode microplate readers, enabling precise timed reagent delivery for kinetic and flash-luminescence assays.
  • Adds two precision injectors for automated reagent delivery.
Catalog No. 50-000-14673 Supplier Molecular Devices Supplier No. ID3INJG
Add to Cart
Add to Cart
Request a Quote
The SpectraMax iD3 Injector System adds automated reagent injection capability to iD3-series multi-mode microplate readers, enabling precise timed reagent delivery for kinetic and flash-luminescence assays.
  • Adds two precision injectors for automated reagent delivery.