Learn More
Molecular Devices Molecular Devices Multi-Mode Microplate Reader, Bottom-Read Luminescence, Injector-Ready
Catalog No. 5000014668
Encompass_Preferred
The SpectraMax iD5 is a compact, five-mode multi-mode microplate reader configurable for bottom-read luminescence and capable of accepting an optional two-injector system for flash assays. It supports 6 to 384-well plates and is suited for dual-luciferase, ATP, and common absorbance and fluorescence workflows.
  • Five built-in read modes: absorbance, fluorescence, luminescence, TRF, and FP.
  • Expandable bottom-read luminescence option for sensitive assays.
  • Optional two-injector SmartInject system with low dead volume and reverse prime.
  • Supports 6 to 384-well plate formats.
  • Injector performance: dispense accuracy ±5% and precision CV ≤2% at 100 μL.
Catalog No. 50-000-14668 Supplier Molecular Devices Supplier No. ID5BLINJ
Add to Cart
Add to Cart
Request a Quote
The SpectraMax iD5 is a compact, five-mode multi-mode microplate reader configurable for bottom-read luminescence and capable of accepting an optional two-injector system for flash assays. It supports 6 to 384-well plates and is suited for dual-luciferase, ATP, and common absorbance and fluorescence workflows.
  • Five built-in read modes: absorbance, fluorescence, luminescence, TRF, and FP.
  • Expandable bottom-read luminescence option for sensitive assays.
  • Optional two-injector SmartInject system with low dead volume and reverse prime.
  • Supports 6 to 384-well plate formats.
  • Injector performance: dispense accuracy ±5% and precision CV ≤2% at 100 μL.