Learn More
Molecular Devices Molecular Devices Multi-Mode Microplate Reader, Five-Mode, Dual Injectors, 6-384-Well
Catalog No. 5000014365
Encompass_Preferred
The SpectraMax iD5 multi-mode microplate reader with dual injectors provides flexible detection for absorbance, fluorescence, luminescence, and time-resolved assays, and enables injector-based kinetic workflows for rapid reagent delivery across plate formats.
  • Five detection modes: absorbance, fluorescence, luminescence, time-resolved fluorescence, fluorescence polarization.
  • Dual injectors with low dead volume and overflow protection for kinetic assays.
  • Supports plate formats from 6 to 384 wells with well scanning up to 20 x 20.
  • Absorbance range 230-1000 nm; fluorescence excitation 250-830 nm, emission 270-850 nm.
  • Temperature control to 66 °C and linear, orbital, and double-orbital shaking.
  • Compatible with data acquisition software and robotic integration for automated workflows.
Catalog No. 50-000-14365 Supplier Molecular Devices Supplier No. ID5INJ
Add to Cart
Add to Cart
Request a Quote
The SpectraMax iD5 multi-mode microplate reader with dual injectors provides flexible detection for absorbance, fluorescence, luminescence, and time-resolved assays, and enables injector-based kinetic workflows for rapid reagent delivery across plate formats.
  • Five detection modes: absorbance, fluorescence, luminescence, time-resolved fluorescence, fluorescence polarization.
  • Dual injectors with low dead volume and overflow protection for kinetic assays.
  • Supports plate formats from 6 to 384 wells with well scanning up to 20 x 20.
  • Absorbance range 230-1000 nm; fluorescence excitation 250-830 nm, emission 270-850 nm.
  • Temperature control to 66 °C and linear, orbital, and double-orbital shaking.
  • Compatible with data acquisition software and robotic integration for automated workflows.