Learn More
Molecular Devices Molecular Devices Membrane Holder, Western blot, For Microplate Reader
Catalog No. 5000014654
Encompass_Preferred
The ScanLater membrane holder secures western blot membranes for scanning within the ScanLater western blot detection workflow on compatible multi-mode microplate readers. It positions membranes for reproducible scanning and integrates with membrane detection modules to enable time-resolved fluorescence measurements.
  • Holds standard western blot membranes securely for scanning.
  • Positions membranes reproducibly to improve scan consistency.
  • Integrates into a microplate reader workflow for membrane detection.
  • Enables time-resolved fluorescence membrane measurements when used with compatible detection hardware.
  • Simple insertion and removal for efficient sample handling.
Catalog No. 50-000-14654 Supplier Molecular Devices Supplier No. 65900146
The ScanLater membrane holder secures western blot membranes for scanning within the ScanLater western blot detection workflow on compatible multi-mode microplate readers. It positions membranes for reproducible scanning and integrates with membrane detection modules to enable time-resolved fluorescence measurements.
  • Holds standard western blot membranes securely for scanning.
  • Positions membranes reproducibly to improve scan consistency.
  • Integrates into a microplate reader workflow for membrane detection.
  • Enables time-resolved fluorescence membrane measurements when used with compatible detection hardware.
  • Simple insertion and removal for efficient sample handling.