Learn More
AdipoGen IDA
SDP
Supplier:  AdipoGen CDXM0001M250
Encompass_Preferred
Chemical. CAS 163556-04-5. Formula C12H9NO4. MW 231.2. Synthetic. Amino compounds having primary groups such as amino acids or aminocyclitol antibiotics lack chromopores or fluorophores in their molecules. Derivatization is required for their detection with high sensitivity using high-performance liquid chromatography or high-performance apillary electrophoresis with photometric and fluorimetric detection. Reagent for the derivatisation of primary amines. The alanine-derivative shows a 3 times stronger fluorescence than o-phthalaldehyde. The reagent has been used in the HPCE-determination of aminoacids.
Catalog No. 50-256-0592