Learn More
Bioworld IDA CHELATING SEPAROPOREA
Supplier:  Bioworld 201810204
Encompass
IDA CHELATING SEPAROPORE (AGAROSE) (METAL-FREE): Metal-Chelating Separopore Affinity matrix for proteins with metal bnding properties with exposed amino acids such as histidine and cysteine.; Can be charged with Zn, Cu, Co and Ni metal i...
Catalog No. 50-197-710