Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Idalopirdine hydrochloride | 467458-02-2 | 100.0% | C20H20ClF5N2O | 10MG
SDP

Catalog No. 5000259710
Encompass_Preferred

Idalopirdine hydrochloride is the hydrochloride salt of idalopirdine, a selective 5-HT6 receptor antagonist supplied as a high-purity research reagent for pharmacology and neuroscience studies. It is used in preclinical investigations of cognition and neurodegenerative disease mechanisms and is available as a solid and as a DMSO solution for assay use.

  • Selective 5-HT6 receptor antagonist suitable for cognition research.
  • High reported purity (99.96%).
  • Available in small milligram quantities and as a 10 mM solution in DMSO.
  • Molecular formula C20H20ClF5N2O and molecular weight 434.83 g/mol.
  • For research use only; not for human consumption.

Catalog No. 50-002-59710 Supplier Medchemexpress LLC Supplier No. HY14338A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Idalopirdine hydrochloride is the hydrochloride salt of idalopirdine, a selective 5-HT6 receptor antagonist supplied as a high-purity research reagent for pharmacology and neuroscience studies. It is used in preclinical investigations of cognition and neurodegenerative disease mechanisms and is available as a solid and as a DMSO solution for assay use.

  • Selective 5-HT6 receptor antagonist suitable for cognition research.
  • High reported purity (99.96%).
  • Available in small milligram quantities and as a 10 mM solution in DMSO.
  • Molecular formula C20H20ClF5N2O and molecular weight 434.83 g/mol.
  • For research use only; not for human consumption.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.