Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Idalopirdine hydrochloride | 467458-02-2 | 99.6% | C20H20ClF5N2O | 1 ML
SDP

Catalog No. 5000354235
Encompass_Preferred

Idalopirdine hydrochloride (Lu AE58054 hydrochloride) is a potent, selective 5-HT6 receptor antagonist with a Ki value of 0.83 nM. It is for research use only and may be used in studies of Alzheimer's disease and schizophrenia, among other related disorders.

  • Potent, selective 5-HT6 receptor antagonist with a Ki value of 0.83 nM
  • May be used in studies of Alzheimer's disease and schizophrenia
  • Soluble in DMSO (≥ 25 mg/mL) and H₂O (2 mg/mL)
  • Solid form storage: 4°C, sealed, away from moisture
  • In solvent storage: -80°C for 6 months; -20°C for 1 month
  • Molecular weight: 434.83
  • Appearance: White to gray solid

Catalog No. 50-003-54235 Supplier Medchemexpress LLC Supplier No. HY14338A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Idalopirdine hydrochloride (Lu AE58054 hydrochloride) is a potent, selective 5-HT6 receptor antagonist with a Ki value of 0.83 nM. It is for research use only and may be used in studies of Alzheimer's disease and schizophrenia, among other related disorders.

  • Potent, selective 5-HT6 receptor antagonist with a Ki value of 0.83 nM
  • May be used in studies of Alzheimer's disease and schizophrenia
  • Soluble in DMSO (≥ 25 mg/mL) and H₂O (2 mg/mL)
  • Solid form storage: 4°C, sealed, away from moisture
  • In solvent storage: -80°C for 6 months; -20°C for 1 month
  • Molecular weight: 434.83
  • Appearance: White to gray solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.