Learn More
Medchemexpress LLC Idalopirdine hydrochloride | 467458-02-2 | 99.6% | C20H20ClF5N2O | 100 MG
SDP
Catalog No. 5000356200
Encompass_Preferred
Idalopirdine hydrochloride is a potent and selective 5-HT6 receptor antagonist with a Ki value of 0.83 nM. It is primarily used in research studies related to Alzheimer's disease and schizophrenia. It has shown to reduce food intake and body weight in over-eating rat models, and enhance cortical gamma oscillations.
  • Potent and selective 5-HT6 receptor antagonist
  • Ki value of 0.83 nM
  • Used in studies of Alzheimer's disease and schizophrenia
  • Reduces food intake and body weight in over-eating rat models
  • Enhances cortical gamma oscillations
  • Suitable for neurological and neurodegenerative disease research
Catalog No. 50-003-56200 Supplier Medchemexpress LLC Supplier No. HY14338A100MG
Idalopirdine hydrochloride is a potent and selective 5-HT6 receptor antagonist with a Ki value of 0.83 nM. It is primarily used in research studies related to Alzheimer's disease and schizophrenia. It has shown to reduce food intake and body weight in over-eating rat models, and enhance cortical gamma oscillations.
  • Potent and selective 5-HT6 receptor antagonist
  • Ki value of 0.83 nM
  • Used in studies of Alzheimer's disease and schizophrenia
  • Reduces food intake and body weight in over-eating rat models
  • Enhances cortical gamma oscillations
  • Suitable for neurological and neurodegenerative disease research