Learn More
Medchemexpress LLC Idalopirdine hydrochloride | 467458-02-2 | 99.6% | C20H20ClF5N2O | 25 MG
SDP
Catalog No. 5000356977
Encompass_Preferred
Idalopirdine hydrochloride (Lu AE58054 hydrochloride) is a potent, selective 5-HT6 receptor antagonist with a Ki value of 0.83 nM. It is intended for research and analytical applications and may be used in studies of Alzheimer's disease and schizophrenia, among other related disorders. This product has been tested and complies with given specifications, but has not been fully validated for medical applications.
  • Potent, selective 5-HT6 receptor antagonist
  • Ki value of 0.83 nM
  • May be used in studies of Alzheimer's disease
  • May be used in studies of schizophrenia
  • Intended for research and analytical applications
Catalog No. 50-003-56977 Supplier Medchemexpress LLC Supplier No. 25MGHY14338A
Idalopirdine hydrochloride (Lu AE58054 hydrochloride) is a potent, selective 5-HT6 receptor antagonist with a Ki value of 0.83 nM. It is intended for research and analytical applications and may be used in studies of Alzheimer's disease and schizophrenia, among other related disorders. This product has been tested and complies with given specifications, but has not been fully validated for medical applications.
  • Potent, selective 5-HT6 receptor antagonist
  • Ki value of 0.83 nM
  • May be used in studies of Alzheimer's disease
  • May be used in studies of schizophrenia
  • Intended for research and analytical applications