Learn More
Medchemexpress LLC Idalopirdine hydrochloride | 467458-02-2 | 99.6% | C20H20ClF5N2O | 50 MG
SDP
Catalog No. 5000354984
Encompass_Preferred
Idalopirdine hydrochloride is a potent and selective 5-HT6 receptor antagonist with a Ki value of 0.83 nM. It is primarily used in research studies related to Alzheimer's disease and schizophrenia. This product is for research use only.
  • Potent and selective 5-HT6 receptor antagonist
  • Ki value: 0.83 nM
  • Supports studies in Alzheimer's disease
  • Supports studies in schizophrenia
  • Reduces food intake and body weight in over-eating rat models
  • Increases gamma power during nPO electrical stimulation
  • Enhances donepezil effects on cortical gamma oscillations
Catalog No. 50-003-54984 Supplier Medchemexpress LLC Supplier No. HY14338A50MG
Idalopirdine hydrochloride is a potent and selective 5-HT6 receptor antagonist with a Ki value of 0.83 nM. It is primarily used in research studies related to Alzheimer's disease and schizophrenia. This product is for research use only.
  • Potent and selective 5-HT6 receptor antagonist
  • Ki value: 0.83 nM
  • Supports studies in Alzheimer's disease
  • Supports studies in schizophrenia
  • Reduces food intake and body weight in over-eating rat models
  • Increases gamma power during nPO electrical stimulation
  • Enhances donepezil effects on cortical gamma oscillations