Learn More
Selleck Chemical LLC Idarubicin HCl 5mg 57852-57-0 4-demethoxydaunorubicin (NSC256439, 4-DMDR) HCl
SDP
Supplier:  Selleck Chemical LLC S12285MG
Encompass_Preferred
Idarubicin HCl (4-demethoxydaunorubicin, NSC256439, 4-DMDR) is a hydrochloride salt form of Idarubicin which is an anthracycline antibiotic and a DNA topoisomerase II (topo II) inhibitor for MCF-7 cells with IC50 of 3.3 ng/mL in a cell-free assay. Idarubicin induces mTOR-dependent cytotoxic autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1049