Learn More
Medchemexpress LLC HY-17381 5mg Medchemexpress, Idarubicin (hydrochloride) CAS:57852-57-0 Purity:>98%
SDP
Catalog No. 501932795
Encompass_Preferred
Medchemexpress, HY-17381 5mg Idarubicin (hydrochloride) CAS:57852-57-0 Idarubicin hydrochloride is an anthracycline antileukemic drug. It inhibits the topoisomerase II interfering with the replication of DNA and RNA transcription. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2795 Supplier Medchemexpress LLC Supplier No. HY173815MG
Medchemexpress, HY-17381 5mg Idarubicin (hydrochloride) CAS:57852-57-0 Idarubicin hydrochloride is an anthracycline antileukemic drug. It inhibits the topoisomerase II interfering with the replication of DNA and RNA transcription. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.