Learn More
TARGETMOL CHEMICALS INC IDARUBICIN HYDROCHLORIDE 25MG
SDP
Catalog No. 502579674
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Idarubicin hydrochloride (Zavedos) a hydrochloride salt form of Idarubicin is a DNA topoisomerase II (topo II) inhibitor for MCF-7 cells ( IC50 3.3 ng/mL). purity: 99%
Catalog No. 50-257-9674 Supplier TARGETMOL CHEMICALS INC Supplier No. T601025MG
Also available in 5 mg 10 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Idarubicin hydrochloride (Zavedos) a hydrochloride salt form of Idarubicin is a DNA topoisomerase II (topo II) inhibitor for MCF-7 cells ( IC50 3.3 ng/mL). purity: 99%