Learn More
Medchemexpress LLC Idarubicin hydrochloride | 57852-57-0 | 99.8% | 533.95 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY17381
Encompass_Preferred
Idarubicin hydrochloride is an anthracycline antileukemic agent that inhibits topoisomerase II, interfering with DNA replication and RNA transcription. It also inhibits the growth of bacteria and yeasts.
  • Anthracycline antileukemic agent
  • Inhibits topoisomerase II
  • Interferes with DNA replication and RNA transcription
  • Inhibits the growth of bacteria and yeasts
  • Shown greater cytotoxicity than daunorubicin or doxorubicin in various in vitro systems
  • Better ability to induce the formation of topoisomerase II-mediated DNA breaks
  • Produces a concentration-dependent reduction in cell growth
  • Produces a concentration-dependent inhibition of DNA synthesis
Catalog No. 50-003-84326