Learn More
Medchemexpress LLC Idarubicin hydrochloride | 57852-57-0 | 99.9% | 533.95 | 1 MG
SDP
Catalog No. 5000384478
Encompass_Preferred
Idarubicin hydrochloride is an anthracycline antileukemic agent. It inhibits topoisomerase II, interfering with DNA replication and RNA transcription. It also inhibits the growth of bacteria and yeasts.
  • Inhibits topoisomerase II
  • Interferes with DNA replication and RNA transcription
  • Inhibits the growth of bacteria and yeasts
  • Greater cytotoxicity than daunorubicin or doxorubicin in various in vitro systems
  • About 57.5-fold and 25-fold more active than doxorubicin and epirubicin, respectively
  • Produces a concentration-dependent reduction in cell growth with an IC50 value of approximately 0.01 μM
  • Produces a concentration-dependent inhibition of DNA synthesis
Catalog No. 50-003-84478 Supplier Medchemexpress LLC Supplier No. HY173811MG
Add to Cart
Add to Cart
Idarubicin hydrochloride is an anthracycline antileukemic agent. It inhibits topoisomerase II, interfering with DNA replication and RNA transcription. It also inhibits the growth of bacteria and yeasts.
  • Inhibits topoisomerase II
  • Interferes with DNA replication and RNA transcription
  • Inhibits the growth of bacteria and yeasts
  • Greater cytotoxicity than daunorubicin or doxorubicin in various in vitro systems
  • About 57.5-fold and 25-fold more active than doxorubicin and epirubicin, respectively
  • Produces a concentration-dependent reduction in cell growth with an IC50 value of approximately 0.01 μM
  • Produces a concentration-dependent inhibition of DNA synthesis