Learn More
Enzo Life Sciences Idarubicin. hydrochloride (5mg). CAS: 57852-57-0
SDP
Supplier:  Enzo Life Sciences ALX380260M005
Encompass_Preferred
More potent, lipophilic and antineoplastic analog of daunorubicin. DNA-damaging effect includes DNA oxidation and methylation, DNA intercalation, inhibition of DNA synthesis, induction of DNA strand breaks and delay of cell cycle progression. Inhibitor of topoisomerase IIα. Produces endonucleolytic cleavage and is a marker of apoptosis. Alternative name: Idamycin, DMDR, 4-Demethoxydaunorubicin. Purity: ≥98% (HPLC). Appearance: Orange crystalline solid. Solubility: Soluble in water or methanol. Long Term Storage: -20°C.
Catalog No. 50-200-8990