Learn More
Selleck Chemical LLC Idasanutlin
SDP
Catalog No. 5000004281
Encompass_Preferred
Idasanutlin (RG7388) is a potent and selective p53-MDM2 inhibitor with IC50 of 6 nM showing improved in vitro binding as well as cellular potency/selectivity
Catalog No. 50-000-04281 Supplier Selleck Chemical LLC Supplier No. S720525MG
Idasanutlin (RG7388) is a potent and selective p53-MDM2 inhibitor with IC50 of 6 nM showing improved in vitro binding as well as cellular potency/selectivity