Learn More
Medchemexpress LLC HY-15676 50mg Medchemexpress, Idasanutlin CAS:1229705-06-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1567650MG
Encompass_Preferred
Medchemexpress, HY-15676 50mg Idasanutlin CAS:1229705-06-9 Idasanutlin (RG7388) is a potent and selective MDM2 antagonist, inhibiting p53-MDM2 binding, with an IC50 of 6 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9155
 Restricted Item