Learn More
Medchemexpress LLC Idasanutlin-d3-1 | 1229705-22-9 | C31H26D3Cl2F2N3O4 | 5 MG
SDP
Catalog No. 5000376094
Encompass_Preferred
Idasanutlin-d3-1 (RG7388-d3-1) is a deuterium-labeled Idasanutlin, a potent antagonist of MDM2/p53, inhibiting relapsed or refractory acute myeloid leukemia. Stable heavy isotopes are incorporated as tracers for quantitation during drug development, and deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-76094 Supplier Medchemexpress LLC Supplier No. HY153939S5MG
Idasanutlin-d3-1 (RG7388-d3-1) is a deuterium-labeled Idasanutlin, a potent antagonist of MDM2/p53, inhibiting relapsed or refractory acute myeloid leukemia. Stable heavy isotopes are incorporated as tracers for quantitation during drug development, and deuteration can affect the pharmacokinetic and metabolic profiles of drugs.