Learn More
Apexbio Technology LLC Idazoxan hydrochloride 79944-56-2 25mg
SDP
Catalog No. 5000088367
Encompass_Preferred
Idazoxan hydrochloride (CAS 79944-56-2) is a selective antagonist of 2-adrenergic and imidazoline receptors that modulates central nervous system signaling by inhibiting endogenous ligand activity at these sites In experimental studies the compound competitively blocks the hypotensive effects of imidazoline agents and has demonstrated efficacy in alleviating motor deficits in Parkinson s disease models Idazoxan hydrochloride is utilized extensively in investigations of neuropharmacological mechanisms and the pathophysiology of neurological disorders
Catalog No. 50-000-88367 Supplier Apexbio Technology LLC Supplier No. B645525
Idazoxan hydrochloride (CAS 79944-56-2) is a selective antagonist of 2-adrenergic and imidazoline receptors that modulates central nervous system signaling by inhibiting endogenous ligand activity at these sites In experimental studies the compound competitively blocks the hypotensive effects of imidazoline agents and has demonstrated efficacy in alleviating motor deficits in Parkinson s disease models Idazoxan hydrochloride is utilized extensively in investigations of neuropharmacological mechanisms and the pathophysiology of neurological disorders